Fura Red potassium salt
Supplier: AAT BIOQUEST INC
|
Warning
|
21045
21047
76483-178EA
437.45
USD
76483-178
76483-180
Fura Red potassium salt
Fura Red potassium salt
Fura Red is a visible light-excitable fura-2 analog that offers unique possibilities for ratiometric measurement of calcium ion in single cells by microphotometry, imaging or flow cytometry when used with single excitation, green-fluorescent calcium indicators.
- Calcium-dependent fluorescence
- Ratiometric detection
- Cell impermeant
Precaución: For research use only
MW:
808.98 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...