Calbryte™ 630 AM
Supplier: AAT BIOQUEST INC
Warning
20720 20721 20722
76484-396EA 249.58 USD
76484-396 76484-398 76484-400
Calbryte™ 630 AM
Calbryte™ 630 AM
Calcium measurement is critical for numerous biological investigations.
  • Improved background/signal ratio
  • Excellent intracellular retention
  • Long-wavelength emission
  • Cell permeant
Precaución: For research use only
ADR: 5,II