Calbryte™ 590 potassium salt
Supplier: AAT BIOQUEST INC
|
Warning
|
20706
20707
76484-392EA
375.27
USD
76484-392
76484-394
Calbryte™ 590 potassium salt
Calbryte™ 590 potassium salt
The intracellular calcium flux assay is a widely used method in monitoring signal transduction pathways and high throughput screening of G protein & 'coupled receptors' (GPCRs) and calcium channel targets.
- Improved background/signal ratio
- Excellent intracellular retention
- Long-wavelength emission
- Cell impermeant
Precaución: For research use only
MW:
1074.98 g/mol |
ADR:
5 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...