Calbryte™ 590 AM
Supplier: AAT BIOQUEST INC
|
Warning
|
20700
20701
20702
76484-386EA
249.58
USD
76484-386
76484-388
76484-390
Calbryte™ 590 AM
Calbryte™ 590 AM
The intracellular calcium flux assay is a widely used method in monitoring signal transduction pathways and high throughput screening of G protein"coupled receptors (GPCRs) and calcium channel targets.
- Improved background/signal ratio
- Excellent intracellular retention
- Long-wavelength emission
- Cell permeant
Precaución: For research use only
MW:
1218.77 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...