Calbryte™ 520 potassium salt
Supplier: AAT BIOQUEST INC
|
Warning
|
20656
20658
76484-366EA
249.58
USD
76484-366
76484-368
Calbryte™ 520 potassium salt
Calbryte™ 520 potassium salt
Calcium measurement is critical for numerous biological investigations.
- Improved background/signal ratio
- Robust
- Cell impermeant
Precaución: For research use only
MW:
909.02 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...