Calbryte™ 520 AM

Supplier: AAT BIOQUEST INC
Warning

20650 20651 20653
76484-360EA 124.31 USD
76484-360 76484-362 76484-364
Calbryte™ 520 AM
Calbryte™ 520 AM

The intracellular calcium flux assay is a widely used method in monitoring signal transduction pathways and high throughput screening of G protein coupled receptors (GPCRs) and calcium channel targets.


  • Improved background/signal ratio
  • Suitable for all major fluorescence platforms
  • Cell permeant


Precaución: For research use only

MW: 1090.9 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR