Cal Red™ R525/650 AM
Supplier: AAT BIOQUEST INC
Warning
20590 20591
76483-088EA 626.51 USD
76483-088 76483-090
Cal Red™ R525/650 AM
Cal Red™ R525/650 AM
The intracellular calcium flux assay is a widely used method for monitoring the activities of GPCRs and calcium channels.
  • Calcium-dependent fluorescence
  • Improved sensitivity
  • Compatible with hts filters
  • Ratiometric detection
  • Cell permeant
Precaución: For research use only
ADR: 5,II