Fluo-4 Pentapotassium Salt

Supplier: AAT BIOQUEST INC
Warning

20555 20556
76483-082EA 375.27 USD
76483-082 76483-084
Fluo-4 Pentapotassium Salt
Fluo-4 Pentapotassium Salt

Calcium measurement is critical for numerous biological investigations.


  • Calcium-dependent fluorescence
  • Steady signal
  • Cell impermeant


Precaución: For research use only

MDL Number: MFCD03456811
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8b3da224f6df79338759b50511bf585d1119977&vid=079f7fbde03dc77e4433d597193b6f6bcbbacab7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR