Cal-590™ potassium salt

Supplier: AAT BIOQUEST INC
Warning

20518 20519
76483-060EA 249.58 USD
76483-060 76483-062
Cal-590™ potassium salt
Cal-590™ potassium salt

Calcium measurement is critical for numerous biological investigations.


  • Improved sensitivity
  • Improved calcium response
  • Long-wavelength emission
  • Cell impermeant


Precaución: For research use only

MW: 1123.03 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR