iFluor® 750 Goat Anti-Rabbit IgG (H+L)
Supplier: AAT BIOQUEST INC
16660 16813
76482-582EA 96.26 USD
76482-582 76482-584
iFluor® 750 Goat Anti-Rabbit IgG (H+L)
Antibodies
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins
Precaución: For research use only.