Sus scrofa (Pig) Recombinant IL10/Interleukin-10 Protein (from E. coli)
Supplier: Sino Biological
62000-WNAE-100 62000-WNAE-20
103666-016EA 704.98 USD
103666-016 103668-890
Sus scrofa (Pig) Recombinant IL10/Interleukin-10 Protein (from E. coli)
Proteins and Peptides
A DNA sequence encoding the porcine IL10 (Q29055) (Ser19-Asn175) was expressed with an initial Met at the N-terminal.