Cole-Parmer® Essentials Heavy-Duty Plastic Bottles, HDPE, Antylia Scientific
Supplier: Antyila Scientific
06025-50 06025-60 06025-70
76534-008PK 141.04 USD
76534-008 76534-010 76534-012
Cole-Parmer® Essentials Heavy-Duty Plastic Bottles, HDPE, Antylia Scientific
Bottles Plastic Bottles
Strong, leakproof storage.
  • Polypropylene closure will not leak
  • Stands up to general storage or shipping
Leakproof and rugged, these high-density polyethylene (HDPE) bottles make excellent storage or shipping containers for liquids or dry chemicals. Bottles are not autoclavable.