PE [R-Phycoerythrin], ReadiUse™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

2500 2501
76484-746EA 124.31 USD
76484-746 76484-748
PE [R-Phycoerythrin], ReadiUse™ fluorescent probe
PE [R-Phycoerythrin]

R-Phycoerythrin (PE) is isolated from red algae.


  • No dialysis required
  • Ready to use
  • High purity

MW: 240000 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b07974981d8e27fb25793068672f898692b77e18&vid=f86f281a2e7971998e6794c60cce9cc339cccdac&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR