Tide Quencher™ 2WS alkyne, Tide Quencher™ fluorescent probe
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
TQ2WS alkyne
2213
2214
76485-020EA
124.31
USD
76485-020
76485-022
Tide Quencher™ 2WS alkyne, Tide Quencher™ fluorescent probe
Tide Quencher™ 2WS alkyne
TQ2WS is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
MW:
527.66 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...