N-Sp-DMAE NHS ester for biomolecular labeling

Supplier: AAT BIOQUEST INC
Warning

26010 26011
76484-994EA 249.58 USD
76484-994 77462-052
N-Sp-DMAE NHS ester for biomolecular labeling
N-Sp-DMAE NHS ester

The acridinium NHS ester can be used to label proteins and nucleic acids.


  • Amine reactive
  • Luminescent detection
  • Sensitive detection

MW: 590.6 g/mol
CAS Number: 194357-64-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69ee5b98674724f46740430e048c0e9de7f52c10&vid=60ae1b77ea824afa383d3cabfb223a8622ca8abc&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=982181f8859430d797c91b2457bb9a5f41d488d7&vid=982181f8859430d797c91b2457bb9a5f41d488d7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR