Phosphoramidite, FAM-xtra™
Supplier: AAT BIOQUEST INC
|
Warning
|
6037
6038
6039
76485-772EA
112.97
USD
76485-772
76485-774
76485-776
Phosphoramidite, FAM-xtra™
Phosphoramidite
There are several ways of labeling an oligonucleotide with fluorescein.
- identical spectra to 6-FAM
- much brighter signal and higher photostability
- nearly pH-independent fluorescence
ADR:
5,II |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)