BOC-Asp(OMe)-FMK

Supplier: AAT BIOQUEST INC

Synonyms: Boc-Asp(OMe)-fluoromethyl ketone, Methyl (S)-3-((tert-butoxycarbonyl)amino)-5-fluoro-4-oxopentanoate

5310 5311
76484-776EA 124.31 USD
76484-776 76484-778
BOC-Asp(OMe)-FMK
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone

The FMK peptides are potent, cell permeable, irreversible capase inhibitors.


  • Key intermediate
  • Cell permeable
  • Multiple applications


For: Molecular biology, Labeling, Proteomics


GHS Symbol: GHS07
Hazard Class 5
Packing Group II
Signalword: Warning

Formula: C₁₁H₁₈FNO₅
MDL Number: MFCD03453073
CAS Number: 187389-53-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c4d71806e906827c7e9e89eaac4654b5c09d550&vid=ba5db17035afe560fd021686e9335196a97eb267&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR