Phalloidin lysine
Supplier: AAT BIOQUEST INC
Warning
5304 5305
76485-748EA 112.97 USD
76485-748 76485-750
Phalloidin lysine
Phalloidin lysine
Phalloidin lysine is a convenient building block that can be used for developing a variety of phalloidin derivatives that may be for studying cellular structures and other biological applications.
  • useful in live or fixed cells
  • high affinity for F-actin
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab0bebd3f41402dd4e7d4a903da71d059ac500fc&vid=ac0dba370ad6322cb7a7df3bf3ccfc4b335dc018&prid=web&clid=SAPDAM