FMOC-Asp(biotinyl-PEG)-OH
Supplier: AAT BIOQUEST INC
Warning
5023 5024
76484-734EA 249.58 USD
76484-734 76484-736
FMOC-Asp(biotinyl-PEG)-OH
FMOC-Asp(biotinyl-PEG)-OH
This PEG biotin-labeled amino acid has excellent solubility in DMF and other solvents used in SPPS.
  • specific reactivity
  • stable conjugation
  • high solubility
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a74991d588a8d5241eca8a0aaf1dad4e591ae4e&vid=5973d07e03e9b332fd5d9e4f97c7f480f99d03f2&prid=web&clid=SAPDAM