N-FMOC-ethylenediamine

Supplier: AAT BIOQUEST INC
Warning

4016 4017
76481-670EA 626.51 USD
76481-670 76481-672
N-FMOC-ethylenediamine
N-FMOC-ethylenediamine

N-FMOC-ethylenediamine is a monoprotected diamine that is widely used as a building block in bioconjugate chemistry.


  • excellent building block
  • multiple applications

Formula: C₁₇H₁₈N₂O₂
MW: 282.34 g/mol
CAS Number: 166410-32-8
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b09e6205cb9cebfdbad824036dc671fda953c5cc&vid=0c52a0ae9adcb951ee59e7ed1e929c41c49925b3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR