6-HEX phosphoramidite for oligonucleotide labeling
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
5'-Hexachlorofluorescein phosphoramidite
6024
6026
76481-836EA
1691
USD
76481-836
76481-838
6-HEX phosphoramidite for oligonucleotide labeling
6-HEX phosphoramidite
There are several ways of labeling an oligonucleotide with fluorescein.
- FRET building block
- Multiple applications
MW:
1050.61 g/mol |
CAS Number:
1360547-55-2 ADR: 5,II |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...