Trypan 0.1 M, Trypan Red Plus™ for dead cell labeling
Supplier: AAT BIOQUEST INC
|
Warning
|
2456
2457
76481-562EA
249.58
USD
76481-562
76481-564
Trypan 0.1 M, Trypan Red Plus™ for dead cell labeling
Trypan
Trypan Red Plus™ is similar to Trypan Blue in cell permeability.
- Minimal cytotoxicity
- High purity
MW:
600 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...