iFluor® 665, Succinimidyl Ester fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

1550 1551 1552 1553
76485-754EA 112.97 USD
76485-754 76485-756 76485-758 76485-760
iFluor® 665, Succinimidyl Ester fluorescent probe
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • pH-insensitive
  • High photostability
  • Brighter signal

ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR