Eu-Cryptate succinimidyl ester, trFluor™ fluorescent probe
Supplier: AAT BIOQUEST INC
|
Warning
|
1430
1431
76484-654EA
375.27
USD
76484-654
76484-656
Eu-Cryptate succinimidyl ester, trFluor™ fluorescent probe
Eu-Cryptate succinimidyl ester
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- no crosslinking
- High stability and selectivity
- Insensitive to fluorescence quenching
- High emission yield
ADR:
5,II |