iFluor® 460, Succinimidyl Ester fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

1022 71022 71502 71552
76485-548EA 237.74 USD
76485-548 76485-550 76485-552 76485-554
iFluor® 460, Succinimidyl Ester fluorescent probe
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR