Cyanine 3.5 alkyne fluorescent probe
Supplier: AAT BIOQUEST INC
|
Warning
|
982
983
76484-704EA
124.31
USD
76484-704
76484-706
Cyanine 3.5 alkyne fluorescent probe
Cyanine 3.5 alkyne
A variety of cyanine 3.5 (Cy3.5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
- High fluorescence quantum yield
- Improved photostability
MW:
1042.09 g/mol |
ADR:
5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...