Cyanine 3.5 azide fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

980 981
76484-700EA 124.31 USD
76484-700 76484-702
Cyanine 3.5 azide fluorescent probe
Cyanine 3.5 azide

A variety of cyanine 3.5 (Cy3.5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.


  • High fluorescence quantum yield
  • Improved photostability

MW: 1073.11 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb60f5624c6a48371e10e1d42a5b7d592208ef19&vid=cb60f5624c6a48371e10e1d42a5b7d592208ef19&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8df97705a6f2e1b10c117eb73a7397d488383b7f&vid=d7b128e7705469db15aa0ec96cb63ec12dc37378&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR