2',3'-cGAMP second messenger

Supplier: AAT BIOQUEST INC
Warning

20310 20311
76485-116EA 582.53 USD
76485-116 76485-118
2',3'-cGAMP second messenger
2',3'-cGAMP

2',3'-cGAMP or cGAMP (Cyclic guanosine monophosphate"adenosine monophosphate) is produced in mammalian cells by cGAS (cGAMP synthase) from ATP and GTP upon cytosolic DNA stimulation.


  • Useful for immunity research
  • Multiple applications

MW: 674.42 g/mol
CAS Number: 1441190-66-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6140abe7675779bcc6399788c7be87745ff158c&vid=f6140abe7675779bcc6399788c7be87745ff158c&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8487368f2c6ff6b4a87432b6aa12879f1053ada&vid=a72b6153a87779407e422fec3806ebe083c5089f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR