EDTA, AM ester metal ion chelator

Supplier: AAT BIOQUEST INC
Warning

19010 19011
76485-132EA 124.31 USD
76485-132 76485-134
EDTA, AM ester metal ion chelator
EDTA, AM ester

Ethylenediaminetetraacetic acid (EDTA) might be the most popular metal ion chelator used for both industrial and medical purposes.


  • Excellent metal chelator
  • Multiple applications

CAS Number: 162303-59-5
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ec4f685e9f6d8fcd7cae9a30a2126881928e46f&vid=6155ffacf55e8a1673beb43ba642717ddca623e5&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abba6649012c71a1365987f5f2c1d9587516e67d&vid=abba6649012c71a1365987f5f2c1d9587516e67d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR