WST-8 50 mM, ReadiUse™ for assessing cell metabolism

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 2-(3-(2-Methoxy-4-nitrophenyl)-2-(4-nitrophenyl)-2H-tetrazol-3-ium-5-yl)-5-sulphobenzenesulphonate, sodium salt, 5-(2,4-Disulphophenyl)-3-(2-methoxy-4-nitrophenyl)-2-(4-nitrophenyl)-2H-tetrazolium inner salt monosodium salt

15705 15706 15707
76484-470EA 96.26 USD
76484-470 76484-472 76484-474
WST-8 50 mM, ReadiUse™ for assessing cell metabolism
WST-8

WST-8 (2-(2-methoxy-4-nitrophenyl)-3-(4-nitrophenyl)-5-(2,4-disulfophenyl)-2H-tetrazolium, monosodium salt) is a water-soluble tetrazolium salt used for assessing cell metabolic activity.


  • Improved sensitivity
  • Water soluble
  • High stability
  • Low cytotoxicity

Formula: C₂₀H₁₃N₆O₁₁S₂·Na
MW: 600.47 g/mol
MDL Number: MFCD09264687
CAS Number: 193149-74-5
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28b7ffb553d50a64cb893185059c58eb13814248&vid=28b7ffb553d50a64cb893185059c58eb13814248&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fbeba4d47f5883428d6a35d6b51a87e0ae37d85&vid=df69d2ca107bba9f8352527730cc5ab502840edb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR