Cyluc1 for in vivo imaging

Supplier: AAT BIOQUEST INC
Warning

Synonyms: (4S)-2-(6,7-Dihydro-5H-thiazolo[4,5-f]indol-2-yl)-4,5-dihydro-thiazole-4-carboxylic acid, (S)-2-(6,7-Dihydro-5H-thiazolo[4,5-f]indol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid, CycLuc1

12480 12481 12482
76484-592EA 124.31 USD
76484-592 76484-594 76484-596
Cyluc1 for in vivo imaging
Cyluc1

Cyluc1 is a synthetic cyclic alkylaminoluciferin that exhibits properties that are superior to those of D-luciferin for in vivo imaging due to its red-shifted luminescence.


  • High-affinity luciferase binding
  • Significantly higher light output
  • 10X brighter signal than D-luciferin

MW: 305.37 g/mol
MDL Number: MFCD28133582
CAS Number: 1247879-16-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ce9ab8abd4e49b9134731913f6403775e4d826d&vid=698626d3b453490cc3400ba4e79e8f3b0a2ff667&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f664571870a5caa6d7fd06a6a9741dde6f4356ff&vid=f664571870a5caa6d7fd06a6a9741dde6f4356ff&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR