Cell Meter™ Colorimetric MTT Cell Proliferation Kit, AAT Bioquest
Supplier: AAT BIOQUEST INC
22768 22769
76485-628EA 358.05 USD
76485-628 76485-630
Cell Meter™ Colorimetric MTT Cell Proliferation Kit, AAT Bioquest
Assays Cellular Assays
Cell Meter™ assay kits are a set of tools for monitoring cell viability.
  • Mix and read format
  • 10x more sensitive
  • Minimal hands-on time necessary
  • No solubilization necessary
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a151e10813b2b7e04a6dddff8286b5c12c7b8c62&vid=25c0698d3a9458826d4fb28c9165d20c776df62d&prid=web&clid=SAPDAM