Amplite® Colorimetric Total NAD and NADH Assay Kit , AAT Bioquest
Supplier: AAT BIOQUEST INC
15258 15275
76482-342EA 375.27 USD
76482-342 76482-370
Amplite® Colorimetric Total NAD and NADH Assay Kit , AAT Bioquest
Assays Enzyme Assays
Nicotinamide adenine dinucleotide (NAD⁺) and nicotinamide adenine dinucleotide phosphate (NADP⁺) are two important cofactors found in cells.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Precaución: For research use only.