Transfectamine™ 5000 Transfection Reagent, AAT Bioquest
Supplier: AAT BIOQUEST INC
Transfectamine™ 5000 Transfection Reagent is a powerful and versatile transfection reagent for the introduction of nucleic acids into eukaryotic cells, or more specifically, into animal cells.
- Easy-to-use, minimal steps required
- High transfection efficiency
- Works with a wide variety of eukaryotic cells
- Effective on dissociated cells and slices
Precaución: For research use only.