Simple Green® Crystal Industrial Cleaner/Degreaser, Essendant
Supplier: Janitorial Supplies
SMP19128 SMP19055 SMP19005
76500-270CT 193.03 USD
76500-270 76500-320 76500-398
Simple Green® Crystal Industrial Cleaner/Degreaser, Essendant
Cleaning Agents Detergents and Surface Cleansers
Simple Green Crystal Industrial Cleaner and Degreaser is a powerful, industrial-strength cleaner and degreaser formulated for critical cleaning applications that demand high performance from a fragrance-free and color-free solution.
  • Powerful, all-purpose cleaning and degreasing with no added color or scent
  • Readily biodegradable formula is VOC compliant, butyl-free, and phosphate-free
  • Non-abrasive, non-corrosive and clean-rinsing formula
  • Safe alternative to caustic cleaners and solvents
  • Cost-effective concentrate can be used manually or with cleaning equipment
  • Chemical compound: water based non-ionic surfactant
This highly-trusted cleaner utilizes a non-flammable and biodegradable formula, making it the ideal cleaning solution for a variety of industries where sanitation and safety are of equal concern. Clean-rinsing, concentrated, and cost-effective, Simple Green Crystal is the sound choice for food processing and handling, industrial, manufacturing, electronics, and parts washing industries. It cuts through tough grease, oils, built-up grime, and other residue without harsh or volatile chemicals. The formula is NSF labeled A1, A2, A4, A8, B1, and C1 for use in commercial kitchens, food processing plants, and other areas where food is prepared or handled. It is able to validate below 10 parts per million in rinsability requirements for cleaning in cosmetics, medical, and pharmaceutical environments where Good Manufacturing Practices (GMP) are followed. OSHA exempt from special handling and personal protection requirements, Crystal Simple Green is a safer alternative to caustic cleaners and solvents.