JM109 Chemically Competent Cells, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
1062-24 1062-48
104013-276PK 171.92 USD
104013-276 104013-278
JM109 Chemically Competent Cells, Intact Genomics
Competent Cells
Intact Genomics JM109 chemically competent Escherichia coli cells are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning.
  • recA– and endA– to minimize recombination and improve the quality of plasmid DNA
  • Contains F’ episome for blue/white screening
JM109 is a K strain that is recA– and endA– to minimize recombination and improve the quality of plasmid DNA. Suitable for high efficiency transformation in a wide variety of applications. Genotype: endA1, recA1, gyrA96, thi, hsdR1More Product Information