CrossLab Internal Standard Kits and Supplies for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
1610132400 9910124100 9910124000
AG1610132400EA 17.74 USD
AG1610132400 AG9910124100 AG9910124000 AG1610126500
CrossLab Internal Standard Kits and Supplies for ICP-OES
Chromatography Supplies Chromatography Spare Parts
Internal standard kit for ICP-OES. Use with organic samples.
  • Extend quality and durability of instruments
  • Reduce sample prep time with on-line addition of internal standard
  • Save up to 15% compared to purchasing supplies individualy
The ICP-OES internal standard kit includes standard PVC pump tubing, EzyFit connectors, five Y pieces and connecting tubing. Use it with aqueous, organic, or high TDS samples to meet your toughest measurement challenges.