Bond Elut Sodium Sulfate Drying Cartridges, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
12131033 12132004 12162052B
AG12131033PK 208 USD
AG12131033 AG12132004 AG12162052B
Bond Elut Sodium Sulfate Drying Cartridges, Agilent Technologies
Chromatography Columns
Bond Elut sodium sulfate drying cartridges simplify drying processes with prepacked devices.
  • Easy to use
  • Available in different formats
  • Prepacked for convenience
Simplify sodium sulfate-mediated drying steps by using cartridges prepacked with ACS grade, granular, anhydrous sodium sulfate. Available in three formats: LRC (large reservoir capacity), Bond Elut Jr, and straight barrels.