SP Bel-Art Magic Touch 2™ Ice Pans, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
M16807-4102 M16807-4104
490014-052EA 222.19 USD
490014-052 490014-054
SP Bel-Art Magic Touch 2™ Ice Pans, Bel-Art Products, a part of SP
Ice Containers
Superior insulating properties combined with high chemical resistance make Magic Touch 2™ Ice Pans compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, salt slurries, dry ice methanol slurries, or liquid nitrogen.
  • Expanded urethane laboratory ice pans are durable and non-sweating
  • Able to withstand a wide temperature range: –196 to 100 °C (–320 to 212 °F)
  • Impervious to moisture and odors
  • Features convenient pour spout, drip-proof rims and fill line indicator
  • Lightly textured non-slip finish and raised edge provides secure transport
Includes snug-fitting lid that stores neatly under base when not in use.
Información para pedidos: Magic Touch 2™ Ice Pans are available in various sizes and colors.