384-Well Plates, Caplugs
Supplier: Caplugs
222-8210-01I 290-8217-010 290-8221-W10 290-8225-Z10
76520-626CT 287.45 USD
76520-626 76520-628 76520-630 76520-632
384-Well Plates, Caplugs
Microplates Deep Well Microplates
These 384-well plates are made of polystyrene, the most commonly used polymer. White plates give the highest possible signal reflection with low background fluorescence. Black plates reduce back scatter light.
  • Distinct alpha-numeric well designations
  • Well volume: 0.112 ml
  • Flat bottom only
All of the lids have raised rings to provide a snug fit, reduce excess evaporation and minimize condensation.