Microcentrifuge Tubes, Caplugs
Supplier: Caplugs
214-3014-020 214-3025-030 214-3027-030 214-3410-080 214-3412-030 214-3439-020 214-3442-080 214-3710-010 214-3711-080 214-3721-010 214-3725-030
76520-604BG 61.12 USD
76520-604 76520-606 76520-608 76520-610 76520-612 76520-614 76520-616 76520-618 76520-620 76520-622 76520-624
Microcentrifuge Tubes, Caplugs
Tubes Microcentrifuge Tubes
Microcentrifuge tubes fit all standard microcentrifuges and cover a range from 0.25 to 1.8 ml.
  • Polypropylene tubes withstand up to 11600×g
  • Polystyrene tubes withstand 6200×g and are useful in lipid fractionation
  • Polyethylene tubes withstand up to 9000×g
Available with or without integrated tethered cap.