12-Well Plates, Caplugs
Supplier: Caplugs
222-8046-01F 333-8026-01F
76520-646CT 307.23 USD
76520-646 76520-648
12-Well Plates, Caplugs
Microplates Deep Well Microplates
12-well plates are gamma-irradiated for sterility and available in both sterile and treated versions.
  • Distinct alpha-numeric well designations
  • Well volume: 6.76 ml
  • Lid included with plate
Available in individually packed units.