CrossLab Detector Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
G4212-68001 G1314-65061
AGG4212-68001KT 1576 USD
AGG4212-68001 AGG1314-65061
CrossLab Detector Kits for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Maintenance kits for your VWD and DAD/MWD.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
Agilent offers maintenance kits to ensure the continued performance of your variable wavelength detector (VWD) and diode array detector/multiple wavelength detector (DAD/MWD). VWD kits include windows, gaskets, screw kits and rings necessary for VWD equipment, and options are available for standard, micro, semi-micro, and high-pressure flow cells. DAD/MWD kits provide a range of options for different capacity flow cells and include all necessary ferrules, seals, wrenches, and torque adaptors required to deliver excellent performance in DAD/MWD applications.