Filtering Flasks, with Plastic Hose Connection, Sartorius
Supplier: Sartorius
16606 16672
76500-754EA 140.6 USD
76500-754 76500-762
Filtering Flasks, with Plastic Hose Connection, Sartorius
Flasks Filter Flasks
These vacuum-resistant filtering flasks made of DURAN® borosilicate glass, can be used in filtrate collection.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard according to DIN 12476.