Anti-Dengue Human Monoclonal Antibody [clone: RV25]
Supplier: ReVacc Scientific
K34Q76-100UG K34Q76-500UG
76515-408EA 332.94 USD
76515-408 76515-410
Anti-Dengue Human Monoclonal Antibody [clone: RV25]
Antibodies
This antibody has been tested to bind to envelope domain III subunit proteins of Dengue virus serotypes 1, 2, 3 and 4.