Anti-MERS Human Monoclonal Antibody [clone: RV45]
Supplier: ReVacc Scientific
B73K78-100UG B73K78-500UG
76515-400EA 332.94 USD
76515-400 76515-402
Anti-MERS Human Monoclonal Antibody [clone: RV45]
Antibodies
This antibody has been tested to bind to trimeric form of MERS-CoV S protein, and show neutralizing activity against MERS pseudovirus (Catalog # T53L53).