Anti-SARS Human Monoclonal Antibody [clone: RV38]
Supplier: ReVacc Scientific
X42K12-100UG X42K12-500UG
76515-396EA 332.94 USD
76515-396 76515-398
Anti-SARS Human Monoclonal Antibody [clone: RV38]
Antibodies
This antibody has been tested to bind to receptor binding domain (RBD) of spike protein of SARS-CoV (2003 strain), and show neutralizing activity against SARS pseudovirus (Catalog # V44X39).