Virus Recombinant SARS-COV-2 Trimeric S2 (from Mammalian Cells)
Supplier: ReVacc Scientific
B85Q65-100UG B85Q65-500UG
76515-448EA 386.23 USD
76515-448 76515-450
Virus Recombinant SARS-COV-2 Trimeric S2 (from Mammalian Cells)
Proteins and Peptides
A recombinant trimeric form of S2 subunit (right downstream of S1/S2 cleavage site from AA# 690) of spike glycoprotein (S protein) of SARS-Cov-2 (GenBank: MN908947) was produced by human embryonic kidney HEK293F cells, followed by purification.
  • Protein function: Bind to its receptor
This product contains ectodomain of S2 protein, a pair of mutations (N1067P and L1068P), a HRV3C cleavage site, T4 foldon trimerization domain, C-terminal 8 hexa-histidine tag and twin-strep tag. Twin-strep tag can be used for streptavidin labeling.
This product has been run on SDS-PAGE with reduced condition and SEC, with results proving trimerization and glycosylation. This product has been tested to bind to our S2 control antibody, RV82 (Catalog # V87W71).