Lanosterol 50%

Supplier: Thermo Scientific Chemicals

46672-0050 46672-0250
TS46672-0050EA 112 USD
TS46672-0050 TS46672-0250
Lanosterol 50%
Lanosterol
Formula: C₃₀H₅₀O
Melting Pt: 137°C
Storage Temperature: Ambient
MDL Number: MFCD00021108
CAS Number: 79-63-0
EINECS: 201-214-9

Order Now

Specification Test Results

Appearance White to light yellow to green powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥ 50.0%
Specific optical rotation +60.0° to +72.0° (20°C, 589nm) (c=1, Toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR