Mephenytoin

Supplier: Thermo Scientific Chemicals
Warning

46669-0250 466691000
TS46669-0250EA 157 USD
TS46669-0250 TS466691000
Mephenytoin
Mephenytoin
Formula: C₁₂H₁₄N₂O₂
MW: 218.25 g/mol
Melting Pt: 135°C…137°C
Storage Temperature: Refrigerator
MDL Number: MFCD00022407
CAS Number: 50-12-4
EINECS: 200-012-8
Merck Index: 13.5878

Order Now

Specification Test Results

Appearance White to almost white crystalline powder
Infrared spectrum Conforms
HPLC ≥ 98.0% (sum of isomers)
Water ≤ 2.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR